Order Entry
Export
Orders LinkContactUsLinkComponent
PYR-41 ≥98% Ubiquitin-activating enzyme (E1) inhibitor

PYR-41 ≥98% Ubiquitin-activating enzyme (E1) inhibitor

Supplier:   Abcam
PYR-41 ≥98% Ubiquitin-activating enzyme (E1) inhibitor
Supplier:  Abcam
Certificates
PYR-41 ≥98% Ubiquitin-activating enzyme (E1) inhibitor

2 Options Available Below

Product Details & Documents

Ubiquitin-activating enzyme (E1) inhibitor (IC₅₀ =5 μM). Inhibits ubiquitination and proteasomal degradation. Additionally increases sumoylation of proteins and induces p53-mediated apoptosis.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Ethyl 4-(4-((5-nitrofuran-2-yl)methylene)-3,5-dioxopyrazolidin-1-yl)benzoate
  • CAS Number:418805-02-4
  • Molecular Weight:371,3 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% Ethyl 4-[(4Z)-4-[(5-nitrofuran-2-yl)methylidene]-3,5-dioxopyrazolidin-1-yl]benzoate
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 175,00 €
    175,00 €
    1 * 5 mg
       175,00 €

  • Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 662,00 €
    662,00 €
    1 * 25 mg
       662,00 €