Order Entry
Export
Orders LinkContactUsLinkComponent
 

Tobramycin free base ≥98% Ribosomal translation inhibitor

Supplier:   Abcam
Tobramycin free base ≥98% Ribosomal translation inhibitor
Supplier:  Abcam
Certificates
Tobramycin free base ≥98% Ribosomal translation inhibitor

2 Options Available Below

Product Details & Documents

Inhibits ribosomal translation. Aminoglycoside antibiotic which binds 30S ribosomal subunit A, preventing functional protein production. Effective against gram-negative bacteria.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  (2S,3R,4S,5S,6R)-4-Amino-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxytetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol
  • CAS Number:32986-56-4
  • Molecular Weight:467,5 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% Tobramycin
Product Family Options

unology General

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 82,00 €
    82,00 €
    1 * 100 mg
       82,00 €

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 308,00 €
    308,00 €
    1 * 500 mg
       308,00 €