Order Entry
Export
Orders LinkContactUsLinkComponent
Luteolin ≥99% anti-inflammatory antioxidant

Luteolin ≥99% anti-inflammatory antioxidant

Supplier:   Abcam
Luteolin ≥99% anti-inflammatory antioxidant
Supplier:  Abcam
Certificates

2 Options Available Below

Product Details & Documents

Antioxidant, anti-inflammatory and anticancer flavonoid. Free radical scavenger and apoptosis inducer. Inhibits TNF-α induced COX-2 expression and supresses PI3K/Akt, NF-κB cell survival pathways.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4-benzopyrone, Natural Yellow 2, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4H-chromen-4-one, 3',4',5,7-Tetrahydroxyflavone
  • CAS Number:491-70-3
  • Molecular Weight:286,24 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-4-benzopyrone
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 92,30 €
    92,30 €
    1 * 10 mg
       92,30 €

  • Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 344,00 €
    344,00 €
    1 * 50 mg
       344,00 €