Paracetamol ≥98% COX-3 inhibitor
Catalog # ABCAAB120990-100
Supplier: Abcam
Jump to:
Product Details & Documents
Selective COX-3 inhibitor (IC₅₀ = 460 μM). NSAID with widely used analgesic and antipyretic effects. Blood-brain barrier permeable.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Synonyms: Acetaminophen, N-Acetyl-4-aminophenol, 4′-Hydroxyacetanilide, 4'-Hydroxyacetanilide, Paracetamolum, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, APAP
- CAS Number:103-90-2
- Molecular Weight:151,16 g/mol
Specifications
Specification Test Results
- Constituents::100% Acetaminophen



