Order Entry
Export
Orders LinkContactUsLinkComponent
Bis-trispropane ≥99% buffer

Bis-trispropane ≥99% buffer

Catalog # ABCAAB146399-50
Supplier:   Abcam
Bis-trispropane ≥99% buffer
Catalog # ABCAAB146399-50
Supplier:  Abcam
CAS Number:  64431-96-5
Bis-trispropane ≥99% buffer
CAS Number:  64431-96-5
Price
97,40 €
Log in for availability
1 * 50 g
Product Details & Documents

Buffer that is useful in the pH 6,3 to 9,5 range. This wide buffering range is due to its two pKa values, 6,8 (pKa1) and 9,0 (pKa2), being so close. Because of the wide buffering range, particularly down to pH 6 to 7, this buffer has been used to enhance the stability or activity of restriction enzymes, compared to Tris buffer (which is a poor buffer below pH 7.5 and has a comparatively large change in pKa with temperature).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  1,3-Bis(tris(hydroxymethyl)methylamino)propane, 1,3-Bis[tris(hydroxymethyl)amino]propane, 2,2'-(Propane-1,3-diylbis(azanediyl))bis(2-(hydroxymethyl)propane-1,3-diol), BIS-TRIS propane, 2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]
  • CAS Number:64431-96-5
  • Molecular Weight:282,34 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Bis-Tris propane