Zinquin ≥95% Zn²⁺ selective fluorescent probe
About this item
Intracellular Zn²⁺ selective fluorescent probe. Forms zinquin-Zn²⁺ complexes in either 1:1 or 2:1 ratios (Kd values are 370 and 850 nM respectively). Zinquin ethyl ester analog. Labels micromolar concentrations of intracellular Zn²⁺ ions with an intense extranuclear fluorescence emitting in the blue range.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:151606-29-0
- Molecular Weight:386,4 g/mol
3 Options Available Below
Product Details & Documents
Intracellular Zn²⁺ selective fluorescent probe. Forms zinquin-Zn²⁺ complexes in either 1:1 or 2:1 ratios (Kd values are 370 and 850 nM respectively). Zinquin ethyl ester analog. Labels micromolar concentrations of intracellular Zn²⁺ ions with an intense extranuclear fluorescence emitting in the blue range.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:151606-29-0
- Molecular Weight:386,4 g/mol
Specifications for Product Family
Specification Test Results
- Constituents::100% Zinquin



