Order Entry
Export
Orders LinkContactUsLinkComponent
Letrozole ≥99% Aromatase inhibitor

Letrozole ≥99% Aromatase inhibitor

Supplier:   Abcam
Letrozole ≥99% Aromatase inhibitor
Supplier:  Abcam
Certificates

2 Options Available Below

Product Details & Documents

Highly potent, reversible, selective aromatase inhibitor (IC₅₀ =11,5 nM). Inhibits estradiol synthesis. Potent antitumor effects in vivo. Cytochrome P450 inhibitor.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  1-Bis(4-cyanophenyl)methyl-1,2,4-triazole, 4,4'-(1,2,4-Triazol-1-ylmethylene)dibenzonitrile
  • CAS Number:112809-51-5
  • Molecular Weight:285,3 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    99% Letrozole
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 82,00 €
    82,00 €
    1 * 10 mg
       82,00 €

  • Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 231,00 €
    231,00 €
    1 * 50 mg
       231,00 €