Order Entry
Export
Orders LinkContactUsLinkComponent
(±)-α-Lipoic acid ≥98% Non-specific free radical scavenger

(±)-α-Lipoic acid ≥98% Non-specific free radical scavenger

Supplier:   Abcam
(±)-α-Lipoic acid ≥98% Non-specific free radical scavenger
Supplier:  Abcam
Certificates
(±)-α-Lipoic acid ≥98% Non-specific free radical scavenger

2 Options Available Below

Product Details & Documents

Non-specific free radical scavenger. Prevents lipid peroxidation and promotes mitochondrial permeability transition in vitro. Anti-oxidant. Active in vivoand in vitro.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)​-​1,2-​Dithiolane-​3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid
  • CAS Number:1077-28-7
  • Molecular Weight:206,3 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    98% D,L-Thioctic acid
Product Family Options

unology General

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 92,30 €
    92,30 €
    1 * 5 g
       92,30 €

  • Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 252,00 €
    252,00 €
    1 * 25 g
       252,00 €