Order Entry
Export
Orders LinkContactUsLinkComponent
Vancomycin hydrochloride ≥95% Tricyclic glycopeptide antibiotic

Vancomycin hydrochloride ≥95% Tricyclic glycopeptide antibiotic

Supplier:   Abcam
Vancomycin hydrochloride ≥95% Tricyclic glycopeptide antibiotic
Supplier:  Abcam
Certificates
Vancomycin hydrochloride ≥95% Tricyclic glycopeptide antibiotic

2 Options Available Below

Product Details & Documents

Tricyclic glycopeptide antibiotic. Inhibits cell wall synthesis in Gram-positive bacteria. Pharmacologically active in vivo.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  (3S,6R,7R,22R,23S,26S,36R,38aR)-3-(2-Amino-2-oxoethyl)-44-[[2-O-(3-amino-2,3,6-trideoxy-3-C-methyl-α-L-lyxo-hexopyranosyl)-β-D-glucopyranosyl]oxy]-10,19-dichloro-2,3,4,5,6,7,23,24,25,26,36,37,38,38a-tetradecahydro-7,22,28,30,32-pentahydroxy-6-[[(2R)-4-methyl-2-(methylamino)-1-oxopentyl]amino]-2,5,24,38,39-pentaoxo-22H-8,11:18,21-Dietheno-23,36-(iminomethano)-13,16:31,35-dimetheno-1H,16H-[1,6,9]oxadiazacyclohexadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine-26-carboxylic acid xhydrochloride, Vancomycin supplement
  • CAS Number:1404-90-6
  • Molecular Weight:1449,2 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% Vancomycin
Product Family Options

unology General

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 76,90 €
    76,90 €
    1 * 250 mg
       76,90 €

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 262,00 €
    262,00 €
    1 * 1 g
       262,00 €