Orders LinkContactUsLinkComponent
Orange G Tracking dye
  ABCAAB146361-25
 :   Abcam
Orange G Tracking dye
  ABCAAB146361-25
 :  Abcam
 :  1936-15-8
Orange G Tracking dye
 :  1936-15-8
76,90 €
1 * 25 g
Azo dye used as a tracking dye for nucleic acids in gel electrophoresis. Used in SDS-capillary electrophoresis as a standard. Also used as a histological stain and as a counterstain for nuclear stains.
 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
 :  Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulphonate, Wool Orange 2G, Orange G sodium salt, CI 16230, Acid Orange 10
  •  :1936-15-8
  •  :452,38 g/mol
  • Constituents::
    100% Dinatrium-7-hydroxy-8-phenylazonaphthalin-1,3-disulfonat