Order Entry
Export
Orders LinkContactUsLinkComponent
Methyl red pH-indicator

Methyl red pH-indicator

Catalog # ABCAAB146368-25
Supplier:   Abcam
Methyl red pH-indicator
Catalog # ABCAAB146368-25
Supplier:  Abcam
CAS Number:  493-52-7
Methyl red pH-indicator
CAS Number:  493-52-7
Price
76,90 €
Log in for availability
1 * 25 g
Product Details & Documents

pH indicator. Azo dye used in the methyl red (MR) test to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose. The transition range is pH 4,4 (red) to pH 6,2 (yellow). Absorption: λmax 410 nm.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS Number:493-52-7
  • Molecular Weight:269,3 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Methyl red