Chloramphenicol ≥99% antibiotic agent
About this item
Broad spectrum antibiotic agent. Binds selectively to A2451 and A2452 residues in the 23S rRNA of the 50S ribosomal subunit. Prevents peptide bond formation and protein chain elongation inhibiting protein biosynthesis. Shows bacteriostatic effects against a wide variety of Gram-positive and Gram-negative bacteria in vivo. Orally active.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:56-75-7
- Molecular Weight:323,13 g/mol
2 Options Available Below
Product Details & Documents
Broad spectrum antibiotic agent. Binds selectively to A2451 and A2452 residues in the 23S rRNA of the 50S ribosomal subunit. Prevents peptide bond formation and protein chain elongation inhibiting protein biosynthesis. Shows bacteriostatic effects against a wide variety of Gram-positive and Gram-negative bacteria in vivo. Orally active.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:56-75-7
- Molecular Weight:323,13 g/mol
Specifications for Product Family
Specification Test Results
- Constituents::99,82% Chloramphenicol
Product Family Options
Product Information
- AvailabilityPrice



