Order Entry
Export
Orders LinkContactUsLinkComponent
Taurine ≥98% Partial glycine receptor agonist

Taurine ≥98% Partial glycine receptor agonist

Catalog # ABCAAB141063-1
Supplier:   Abcam
Taurine ≥98% Partial glycine receptor agonist
Catalog # ABCAAB141063-1
Supplier:  Abcam
CAS Number:  107-35-7
Taurine ≥98% Partial glycine receptor agonist
CAS Number:  107-35-7
Price
71,80 €
Log in for availability
1 * 1 g
Product Details & Documents

Partial glycine receptor agonist. Abundant free amino acid in the retina and central brain and elicits a variety of biological effects. Able to cross the blood-brain barrier.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  2-Aminoethanesulphonic acid
  • CAS Number:107-35-7
  • Molecular Weight:125,15 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Taurine