Order Entry
Export
Orders LinkContactUsLinkComponent
Trypsin Antigen Retrieval Solution, Abcam

Trypsin Antigen Retrieval Solution, Abcam

Trypsin Antigen Retrieval Solution, 25 ml
Catalog # ABCAAB970-25
Supplier:   Abcam
Trypsin Antigen Retrieval Solution, Abcam
Catalog # ABCAAB970-25
Supplier:  Abcam
CAS Number:  
Trypsin Antigen Retrieval Solution, Abcam

Trypsin Antigen Retrieval Solution, 25 ml

CAS Number:  
Price
287,00 €
Log in for availability
1 * 25 mL
Product Details & Documents

Trypsin-based antigen retrieval solution for Protease-Induced Epitope Retrieval (PIER) (ab970).

  • Preservative: 0.05% Sodium azide Constituents: Carrier protein, PBS

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    Trypsin Antigen Retrieval Solution
  • Size:
    25 ml
  • Cat. No.:
    ABCAAB970-25
  • Storage temperature:
    Store at +4°C.

Specifications