Order Entry
Export
Orders LinkContactUsLinkComponent
AEC Substrate Kit, Abcam

AEC Substrate Kit, Abcam

AEC Substrate Kit
Catalog # ABCAAB64252-125
Supplier:   Abcam
AEC Substrate Kit, Abcam
Catalog # ABCAAB64252-125
Supplier:  Abcam
CAS Number:  
AEC Substrate Kit, Abcam

AEC Substrate Kit

CAS Number:  
Price
339,00 €
Log in for availability
1 * 125 mL
Product Details & Documents

AEC substrate kit for use in IHC using HRP / peroxidase detection.

  • pH: 4.7 Constituents: 3% Dimethyl formamide, 0.1% 3-amino-9-ethyl carbazole, 0.03% Hydrogen peroxide

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    AEC Substrate Kit
  • Cat. No.:
    ABCAAB64252-125
  • Storage temperature:
    Store at +4°C.

Specifications