Order Entry
Export
Orders LinkContactUsLinkComponent
10x EDTA Buffer pH 8.0, Abcam

10x EDTA Buffer pH 8.0, Abcam

10x EDTA Buffer pH 8.0
Catalog # ABCAAB64216-125
Supplier:   Abcam
10x EDTA Buffer pH 8.0, Abcam
Catalog # ABCAAB64216-125
Supplier:  Abcam
CAS Number:  
10x EDTA Buffer pH 8.0, Abcam

10x EDTA Buffer pH 8.0

CAS Number:  
Price
139,00 €
Log in for availability
1 * 125 mL
Product Details & Documents

EDTA buffer pH 8.0 for heat-induced antigen retrieval (HIER) during IHC (ab64216).

  • pH: 7.7 - 8.1 Constituents: 0.29% EDTA

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    10x EDTA Buffer pH 8.0
  • Cat. No.:
    ABCAAB64216-125
  • Storage temperature:
    Store at +4°C.

Specifications