Order Entry
Export
Orders LinkContactUsLinkComponent
Multipurpose Mini Spin Columns, Abcam

Multipurpose Mini Spin Columns, Abcam

Multipurpose Mini Spin Columns
Catalog # ABCAAB286861-50
Supplier:   Abcam
Multipurpose Mini Spin Columns, Abcam
Catalog # ABCAAB286861-50
Supplier:  Abcam
CAS Number:  
Multipurpose Mini Spin Columns, Abcam

Multipurpose Mini Spin Columns

CAS Number:  
Price
159,00 €
Log in for availability
1 * 50 items
Product Details & Documents

This product is manufactured by BioVision, an Abcam company and was previously called 6572 Multipurpose Mini Spin Columns.

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    Multipurpose Mini Spin Columns
  • Cat. No.:
    ABCAAB286861-50
  • Storage temperature:
    Store at Ambient.

Specifications