Order Entry
Export
Orders LinkContactUsLinkComponent
Density Gradient Medium (Iodixanol)

Density Gradient Medium (Iodixanol)

Supplier:   Abcam
Density Gradient Medium (Iodixanol)
Supplier:  Abcam
Certificates

2 Options Available Below

Product Details & Documents

This product is manufactured by BioVision, an Abcam company and was previously called M1248 OptiPrep™ Density Gradient Medium.

  • Constituents: 60% Iodixanol

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications for Product Family

Specifications

  • Catalog No:
  • ABCAAB286850-250
  • ABCAAB286850-100
  • Storage conditions:
  • Store at Ambient.
  • Store at Ambient.

Specifications

Product Family Options

unology General

Product Information

  • Availability
    Price
  • ABCAAB286850-250
    Density Gradient Medium (Iodixanol)
    Density Gradient Medium (Iodixanol)
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 344,00 €
    344,00 €
    1 * 250 mL
       344,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB286850-250
    Storage conditions
    Store at Ambient.
  • ABCAAB286850-100
    Density Gradient Medium (Iodixanol)
    Density Gradient Medium (Iodixanol)
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 231,00 €
    231,00 €
    1 * 100 mL
       231,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB286850-100
    Storage conditions
    Store at Ambient.