Order Entry
Export
Orders LinkContactUsLinkComponent
Extraction Enhancer Buffer 50X, Abcam

Extraction Enhancer Buffer 50X, Abcam

Supplier:   Abcam
Extraction Enhancer Buffer 50X, Abcam
Supplier:  Abcam
Certificates

2 Options Available Below

Product Details & Documents

Abcam's Extraction Enhancer Buffer 50X is formulated for the preparation of cell or tissues extracts for use in immunoassays.

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ABCAAB193971-5
  • ABCAAB193971-1
  • Storage temperature:
  • Store at +4°C.
  • Store at +4°C.

Specifications

Product Family Options

Product Information

  • Availability
    Price
  • ABCAAB193971-5
    Extraction Enhancer Buffer 50X
    Extraction Enhancer Buffer 50X
    Log in for availability
    Log in for availability
    1 * 5 mL
    1 * 5 mL - 46,20 €
    46,20 €
    1 * 5 mL
       46,20 €

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB193971-5
    Storage temperature
    Store at +4°C.

unology General

Product Information

  • Availability
    Price
  • ABCAAB193971-1
    Extraction Enhancer Buffer 50X
    Extraction Enhancer Buffer 50X
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 41,00 €
    41,00 €
    1 * 1 mL
       41,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB193971-1
    Storage temperature
    Store at +4°C.