Order Entry
Export
Orders LinkContactUsLinkComponent
Lauryl maltoside solution 10%

Lauryl maltoside solution 10%

Catalog # ABCAAB109857-1
Price
66,70 €
Log in for availability
1 * 1 mL
Product Details & Documents
Synonyms:  DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside
  • CAS Number:69227-93-6