Order Entry
Export
Orders LinkContactUsLinkComponent
Opti-Klear™ Live Cell Imaging Buffer 1X, Abcam

Opti-Klear™ Live Cell Imaging Buffer 1X, Abcam

Opti-Klear™ Live Cell Imaging Buffer 1X
Catalog # ABCAAB275938-60
Supplier:   Abcam
Opti-Klear™ Live Cell Imaging Buffer 1X, Abcam
Catalog # ABCAAB275938-60
Supplier:  Abcam
CAS Number:  
Opti-Klear™ Live Cell Imaging Buffer 1X, Abcam

Opti-Klear™ Live Cell Imaging Buffer 1X

CAS Number:  
Price
129,00 €
Log in for availability
1 * 60 mL
Product Details & Documents

Hepes based imaging buffer formulated to lower background fluorescence and maintain proper pH and osmolarity for hours at 37°C. ab275938 provides an energy source and preserves fluorescent signals.

  • Constituents: 99% HEPES

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    Opti-Klear™ Live Cell Imaging Buffer 1X
  • Cat. No.:
    ABCAAB275938-60
  • Storage temperature:
    Store at +4°C.

Specifications