Order Entry
Export
Orders LinkContactUsLinkComponent
Tissue Clearing Permeabilization Buffer - hydrophobic, Abcam

Tissue Clearing Permeabilization Buffer - hydrophobic, Abcam

Supplier:   Abcam
Tissue Clearing Permeabilization Buffer - hydrophobic, Abcam
Supplier:  Abcam
Certificates
Tissue Clearing Permeabilization Buffer - hydrophobic, Abcam

2 Options Available Below

Product Details & Documents

Tissue Clearing Permeabilization Buffer ab243307 is part of our range of tissue clearing reagents and products that enable you to set up tissue clearing easily and quickly.

  • Constituents: 94% Water, 6% Sodium lauryl sulfate

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ABCAAB243307-100
  • ABCAAB243307-30
  • Storage temperature:
  • Store at Ambient.
  • Store at Ambient.

Specifications

Product Family Options

Product Information

  • Availability
    Price
  • ABCAAB243307-100
    Tissue Clearing Permeabilization Buffer - hydrophobic
    Tissue Clearing Permeabilization Buffer - hydrophobic
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 185,00 €
    185,00 €
    1 * 100 mL
       185,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB243307-100
    Storage temperature
    Store at Ambient.
  • ABCAAB243307-30
    Tissue Clearing Permeabilization Buffer - hydrophobic
    Tissue Clearing Permeabilization Buffer - hydrophobic
    Log in for availability
    Log in for availability
    1 * 30 mL
    1 * 30 mL - 103,00 €
    103,00 €
    1 * 30 mL
       103,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB243307-30
    Storage temperature
    Store at Ambient.