Trypan blue solution 0,4%
About this item
Trypan Blue 0.4% Solution is commonly used as a cell stain to measure cell viability using dye exclusion method. The dye exclusion method is based on the principle that cell impermeable dyes like Trypan blue will stain only dead cells where as viable cells will not be stained.
- Solution is made in PBS, pH 7.4 ± 0.1 Sterile filtered Store at ambient temperature
Trypan Blue 0.4% Solution is routinely used for counting cells with hemocytometer during subculturing of cells. Trypan blue stain can be used to check cell viability accurately and quickly anytime.
- CAS Number:72-57-1
- Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
- MDL Number:MFCD00003969
- Molecular Weight:960.82 g/mol
2 Options Available Below
Product Details & Documents
Trypan Blue 0.4% Solution is commonly used as a cell stain to measure cell viability using dye exclusion method. The dye exclusion method is based on the principle that cell impermeable dyes like Trypan blue will stain only dead cells where as viable cells will not be stained.
- Solution is made in PBS, pH 7.4 ± 0.1 Sterile filtered Store at ambient temperature
Trypan Blue 0.4% Solution is routinely used for counting cells with hemocytometer during subculturing of cells. Trypan blue stain can be used to check cell viability accurately and quickly anytime.
- CAS Number:72-57-1
- Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
- MDL Number:MFCD00003969
- Molecular Weight:960.82 g/mol



