Order Entry
Export
ContactUsLinkComponent
 
VWR® Semi-Quantitative Chlorine Test
Catalog # 92520.600
Supplier:  VWR Chemicals
New Product
VWR® Semi-Quantitative Chlorine Test
Catalog # 92520.600
Supplier:  VWR Chemicals
New Product

Specifications

  • Description:
    Chlorine semi-quantitative test strips

Specifications

About this item

The white paper turns blue-violet in the presence of chlorine.

  • Range: Chlorine (total) - 10, 50, 100 and 200 mg/L Cl₂

    Shelf life: 2 Year(s)

    Storage temperature: 4 to 30 °C (39 to 86 °F)

    Size: 1 reel of 5 m length and 10 mm width

The test is particularly suitable for high range applications in disinfectant solutions which are for example used in food and beverage industry and animal farms.