Nuclear Fast Red solution (Kernechtrot)
Supplier: G-Biosciences
Jump to:
About this item
Conventional stain used for numerous histology and cytology procedures.
- Enhanced stability formula ensures long shelf-life and consistent results
Ready to use formulation of 0.1% nuclear fast red in 5% aluminum sulfate, optimized for histology and cytology procedures where reproducible results are important.
Synonyms: CI 60760, Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonate, Helio Fast Rubin BBL
- CAS Number:6409-77-4
- Formula:C₁₄H₈NNaO₇S
- MDL Number:MFCD00078493
- Molecular Weight:357.28 g/mol
3 Options Available Below
Product Details & Documents
Conventional stain used for numerous histology and cytology procedures.
- Enhanced stability formula ensures long shelf-life and consistent results
Ready to use formulation of 0.1% nuclear fast red in 5% aluminum sulfate, optimized for histology and cytology procedures where reproducible results are important.
Synonyms: CI 60760, Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonate, Helio Fast Rubin BBL
- CAS Number:6409-77-4
- Formula:C₁₄H₈NNaO₇S
- MDL Number:MFCD00078493
- Molecular Weight:357.28 g/mol



