Order Entry
Export
Orders LinkContactUsLinkComponent
Hematoxylin modified solution acc. Mayer

Hematoxylin modified solution acc. Mayer

Hematoxylin modified solution acc. Mayer
Supplier:  G-Biosciences
Certificates
Hematoxylin modified solution acc. Mayer

3 Options Available Below

Product Details & Documents

Core component of Hematoxylin and Eosin (H&E) staining in pathology, research, and diagnostic laboratories.

  • Manufactured with BSC certified reagents

Mayer’s Hemotoxylin Stain, modified for enhanced shelf-life while avoiding use of Chloral Hydrate.

Synonyms:  DcH-48, Natural Black I, Hydroxybrasilin
  • CAS Number:517-28-2
  • Formula:C₁₆H₁₄O₆
  • MDL Number:MFCD00078111
  • Molecular Weight:302.28 g/mol

Documents

Product Family Options

Product Information