Order Entry
Export
Orders LinkContactUsLinkComponent
 

Giemsa's Stain stock solution

Supplier:   G-Biosciences
Giemsa's Stain stock solution
Supplier:  G-Biosciences
Certificates
Giemsa's Stain stock solution

3 Options Available Below

Product Details & Documents

Conventional stain used for Hematoxylin and Eosin (H&E) staining in histology and cytology procedures. Ready to use formulation optimized for histology and cytology procedures where reproducible results are important.

  • This stock solution has been formulated for consistent staining and extended shelf-life

Classic differential stain with a broad spectrum of applications that include cell viability, staining metaphases to look for chromosomal aberrations, blood smears, as well as other histology/cytology applications. Use Giemsa’s azure eosin methylene blue solution for any of its classical applications with minimal troubleshooting due to its standardized recipe.

Synonyms:  Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue
  • CAS Number:51811-82-6
  • MDL Number:MFCD00081642
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 30,90 €
    30,90 €
    1 * 100 mL
       30,90 €

  • Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 90,40 €
    90,40 €
    1 * 500 mL
       90,40 €

  • Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 148,00 €
    148,00 €
    1 * 1 L
       148,00 €