Order Entry
Export
ContactUsLinkComponent
 
VWR® DOSATEST® Test Strips, Nitrite 25
Catalog # 92538.601
Supplier:  VWR Chemicals
New Product
VWR® DOSATEST® Test Strips, Nitrite 25
Catalog # 92538.601
Supplier:  VWR Chemicals
New Product

Specifications

  • Description:
    Semi-quantitative test strips, single parameter kit type

Specifications

About this item

Dosatest® Nitrite 25 test strip enables fast and precise determination of nitrite in liquids.

Ideally suited for the food industry, it covers a measurement range of 0,5 to 25 mg per litre. Results are available within 60 seconds. Especially suitable for applications like whey or spinach, it provides reliable support in quality control.

Parameter: Nitrite

Test method: Test strips

Range: Nitrite - 0,5 - 1 - 2 - 5 - 10 - 25 mg/L NO₂⁻

Shelf life: Two years

Storage temperature: 4 to 30 °C (39 to 86 °F)

Size: 100 test strips in a tube, instruction leaflet