Order Entry
Export
ContactUsLinkComponent
VWR® DOSATEST® Test Strips, Carbonate Hardness
VWR® DOSATEST® Test Strips, Carbonate Hardness
Catalog # 92531.601
Supplier:  VWR Chemicals
New Product
VWR® DOSATEST® Test Strips, Carbonate Hardness
Catalog # 92531.601
Supplier:  VWR Chemicals
New Product

Specifications

  • Description:
    Semi-quantitative test strips, single parameter kit type

Specifications

About this item

Semi-quantitative test strips for the determination of Carbonate hardness.

Ideal for rapid and easy analysis of Carbonate hardness directly on-site. No maintenance or calibration is required.

Parameter: Hardness;Calcium (hardness)

Test method: Test strips

Range: Carbonate hardness - 0, 4 , 8 , 13, 19, 25 °e

Shelf life: Two and half years

Storage temperature: 4 to 30 °C (39 to 86 °F)

Size: 100 test strips in a tube, instruction leaflet