Dr. Ehrenstorfer™ Food Standards, Apigenin
Catalog # EHERDRE-C10290600
Supplier: EHRENSTORFER
New Product
CAS Number:
Specifications
- CAS No.:520-36-5
- Description:Apigenin
- Storage conditions:20 °C
- Formula:C₁₅H₁₀O₅
Specifications
About this item
This RM is intended for use in a laboratory as a calibration and quality control standard or in method development for analytical techniques.
Certifications:
This reference material (RM) was designed, produced and verified in accordance with a registered quality management system ISO 9001. All measurements were performed according to ISO/IEC 17025 by a DAkkS accredited laboratory.