Order Entry
Export
ContactUsLinkComponent
 
NANOCOLOR® Photometric Tube Test Kit for Peroxide
  MANA985871
 :  
undefined
NANOCOLOR® Photometric Tube Test Kit for Peroxide
  MANA985871
 :  

 

  • Chemical method:
    Peroxidase
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Peroxide 2, test no. 8-71
  • Dimensions:
    160×110×119 mm (including packaging)
  • Model:
    MANA985871
  • Platform:
    NANOCOLOR® tube tests
  • Storage temperature:
    2…8 °C
  • Shelf life (months):
    12
  • Weight:
    359.7 g (including packaging)
  • For unit:
    UV/VIS II; VIS II; 500 D; PF-12Plus; Advance
  • Tests per kit:
    19
  • Range:
    0,03 - 2,00 mg/L (H₂O₂)

 

 

Photometric test kit for the determination of peroxide in water samples.

  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Coloured pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting

The test kit is evaluated in 16 mm (OD) tubes. It is preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.

Sea water analysis: Yes
Hazardous material: No
Remark: Measuring range on NANOCOLOR VIS II

 : Rugged box with 19 test tubes.