NANOCOLOR® Photometric Tube Tests, Surfactants
Supplier: MACHEREY-NAGEL
Certificates
About this item
Photometric test kits for the determination of surfactants in water samples.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
- Precisely pre-dosed reagents with constant high quality from batch to batch
- Colored pictogram instructions and easy handling of the tubes
- 16 mm (OD) tubes for easy pipetting
The test kits are evaluated in 16 mm (AD) round cuvettes. They are pre-programmed on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.
Sludge reagent set: No
Digestion set: No
NanOx Metal: No
NanOx N: No
Delivery: Remark: Measuring range on NANOCOLOR VIS II
Specifications
- Model:MANA985032
- Storage temperature:15…25 °C
- Shelf life (months):24
- Weight:494,7 g (including packaging)
Specifications
Related Information
Certificates
Search by Lot NumberAnionic Surfactants 4 Tube Test.pdf NANOCOLOR® Cationic Surfactants, Photometric Tube Tests.pdf Nonionic Surfactants 15 Tube Test.pdf
Additional Product Details
IFU Anionic surfactants 4 Tube Test.pdf IFU NANOCOLOR® Photometric Tube Tests, Cationic Surfactants.pdf IFU Nonionic surfactants 15 Tube Test.pdf