Human Recombinant MOCS2 (from E. coli)
Recombinant Human MOCS2 protein is a Human Full Length protein, in the 1 to 188 aa range, expressed in E. coli, with >90% purity and suitable for SDS-PAGE, MS.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- Form:Liquid
- Protein/peptide name:MOCS2
- Protein synonyms:MCBPE|MOCO1|MOCS2|Molybdopterin synthase catalytic subunit|MOCO1-B|Molybdenum cofactor synthesis protein 2 large subunit|Molybdenum cofactor synthesis protein 2B|Molybdopterin-synthase large subunit|MOCS2B|MPT synthase large subunit
- Purity:>90%
- Concentration:0,25 mg/ml
- Source:E. coli
- Biological activity:Standard
- Shipping temperature:20
- Cat. No.:ABCAAB134527-20
- Protein/peptide type:Recombinant
- Storage conditions:Store at −20 °C
- Formulation:pH: 8. Constituents: 50% Glycerol (glycerin, glycerine), 0,88% Sodium chloride, 0,32% Tris HCl, 0,02% (R*,R*)-1,4-Dimercaptobutan-2,3-diol.
- Conjugation:Unconjugated
- Tested applications:MS, SDS-PAGE
- Species:Human
- Animal-free:N



