Order Entry
Export
Orders LinkContactUsLinkComponent
Human Recombinant PHLPP2 (from E. coli)

Human Recombinant PHLPP2 (from E. coli)

Catalog # ABCAAB125556-20
Supplier:   Abcam
Human Recombinant PHLPP2 (from E. coli)
Catalog # ABCAAB125556-20
Supplier:  Abcam
CAS Number:  
Human Recombinant PHLPP2 (from E. coli)
Human Recombinant PHLPP2 (from E. coli)
CAS Number:  
Price
441,00 €
Log in for availability
1 * 20 µG
Product Details & Documents

Recombinant Human PHLPP2 protein is a Human Fragment protein, in the 766 to 1043 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, WB.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Form:
    Liquid
  • Protein/peptide name:
    PHLPP2
  • Protein synonyms:
    KIAA0931|PHLPPL|PHLPP2|PH domain leucine-rich repeat-containing protein phosphatase 2|PH domain leucine-rich repeat-containing protein phosphatase-like|PHLPP-like
  • Purity:
    >95%
  • Concentration:
    0,1 mg/ml
  • Source:
    E. coli
  • Biological activity:
    Standard
  • Shipping temperature:
    Dry Ice
  • Cat. No.:
    ABCAAB125556-20
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −80 °C
  • Formulation:
    pH: 7,5, Constituents: 25% Glycerol (glycerin, glycerine), 0,88% Sodium chloride, 0,79% Tris HCl, 0,31% Glutathione, 0,004% (R*,R*)-1,4-Dimercaptobutan-2,3-diol, 0,003% EDTA, 0,002% PMSF
  • Conjugation:
    Unconjugated
  • Tested applications:
    SDS-PAGE, WB
  • Species:
    Human
  • Animal-free:
    N

Specifications