Order Entry
Export
Orders LinkContactUsLinkComponent
Human Recombinant KAT2A/GCN5 (from E. coli)

Human Recombinant KAT2A/GCN5 (from E. coli)

Catalog # ABCAAB198113-100
Supplier:   Abcam
Human Recombinant KAT2A/GCN5 (from E. coli)
Catalog # ABCAAB198113-100
Supplier:  Abcam
CAS Number:  
Human Recombinant KAT2A/GCN5 (from E. coli)
Human Recombinant KAT2A/GCN5 (from E. coli)
CAS Number:  
Price
1 220,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Recombinant Human KAT2A/GCN5 protein (Tagged-His Tag) is a Human Fragment protein, in the 727 to 837 aa range, expressed in Escherichia coli, with >99% purity and suitable for SDS-PAGE.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Form:
    Liquid
  • Protein/peptide name:
    KAT2A / GCN5
  • Protein synonyms:
    GCN5|GCN5L2|KAT2A|Histone acetyltransferase KAT2A|General control of amino acid synthesis protein 5-like 2|Histone acetyltransferase GCN5|Histone glutaryltransferase KAT2A|Histone succinyltransferase KAT2A|Lysine acetyltransferase 2A|STAF97|hGCN5
  • Purity:
    >99%
  • Concentration:
    2,13 mg/ml
  • Source:
    E. coli
  • Biological activity:
    Standard
  • Shipping temperature:
    Dry Ice
  • Cat. No.:
    ABCAAB198113-100
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −80 °C
  • Formulation:
    pH: 8, Constituents: 20% Glycerol (glycerin, glycerine), 0,64% Sodium chloride, 0,63% Tris HCl, 0,04% Sorbitan monolaurate, ethoxylated, 0,02% Potassium chloride
  • Conjugation:
    Unconjugated
  • Tested applications:
    SDS-PAGE
  • Species:
    Human
  • Animal-free:
    N

Specifications