Human Recombinant PRPF31 (from Wheat germ)
Catalog # ABCAAB131853-10
Supplier: Abcam
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Wheat germ
- Species:Human
- Seq:1106
- Storage conditions:Store at –80 °C
- Biological activity:Standard
- Form:Liquid
- Animal-free:N
- Protein synonyms:PRP31|PRPF31|U4/U6 small nuclear ribonucleoprotein Prp31|Pre-mRNA-processing factor 31|Serologically defined breast cancer antigen NY-BR-99|U4/U6 snRNP 61 kDa protein|Protein 61K|hPrp31
- Protein/peptide name:PRPF31
- Formulation:pH: 8, Constituents: 0,79% Tris HCl, 0,31% Glutathione
- Shipping temperature:Dry Ice
- Tested applications:ELISA, SDS-PAGE, WB
Specifications
About this item
Recombinant human PRPF31 protein is a human full length protein, in the 1 to 499 aa range, expressed in Wheat germ and suitable for SDS-PAGE, ELISA, WB.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.