Human Recombinant CERT (from E. coli)
Supplier: Abcam
Certificates
About this item
Recombinant Human CERT protein is a Human Fragment protein, in the 347 to 598 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, MS.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C
- Biological activity:Standard
- Form:Liquid
- Animal-free:N
- Protein synonyms:COL4A3BP|STARD11|CERT1|Ceramide transfer protein|hCollagen type IV alpha-3-binding protein|Goodpasture antigen-binding protein|START domain-containing protein 11|StAR-related lipid transfer protein 11|GPBP
- Protein/peptide name:CERT
- Purity:>95%
- Concentration:0,5 mg/ml
- Formulation:pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl
- Shipping temperature:20
- Tested applications:MS, SDS-PAGE