Order Entry
Export
Orders LinkContactUsLinkComponent
Human Native PF4

Human Native PF4

Catalog # ABCAAB81754-100
Supplier:   Abcam
Human Native PF4
Catalog # ABCAAB81754-100
Supplier:  Abcam
CAS Number:  
Human Native PF4
Human Native PF4
CAS Number:  
Price
703,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Native human PF4 protein is a human full length platelet factor 4 protein, in the 32 to 101 aa range with >95% purity and suitable for SDS-PAGE. The predicted molecular weight of ab81754 native protein is 7,8 kDa. Save time and ensure accurate results use our PF4 protein Heparin-neutralizing activity verified by clotting assay. Marker of platelet activation. Optimal protein bioactivity and stability.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Form:
    Liquid
  • Protein/peptide name:
    PF4
  • Protein synonyms:
    CXCL4|SCYB4|PF4|Platelet factor 4|PF-4|C-X-C motif chemokine 4|Iroplact|Oncostatin-A
  • Purity:
    >95%
  • Concentration:
    1,4 mg/ml
  • Shipping temperature:
    Dry Ice
  • Cat. No.:
    ABCAAB81754-100
  • Protein/peptide type:
    Native
  • Storage conditions:
    Store at –80 °C
  • Formulation:
    pH: 7,4, Constituents: 11,6% Sodium chloride, 0,595% HEPES
  • Conjugation:
    Unconjugated
  • Tested applications:
    SDS-PAGE
  • Species:
    Human
  • Animal-free:
    N

Specifications