Order Entry
Export
Orders LinkContactUsLinkComponent
Human Recombinant SLUG (from E. coli)

Human Recombinant SLUG (from E. coli)

Catalog # ABCAAB201878-100
Supplier:   Abcam
Human Recombinant SLUG (from E. coli)
Catalog # ABCAAB201878-100
Supplier:  Abcam
CAS Number:  
Human Recombinant SLUG (from E. coli)
Human Recombinant SLUG (from E. coli)
CAS Number:  
Price
636,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Recombinant Human SLUG protein is a Human Full Length protein, in the 1 to 268 aa range, expressed in Escherichia coli, with >80% purity and suitable for SDS-PAGE.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Form:
    Liquid
  • Protein/peptide name:
    SLUG
  • Protein synonyms:
    SLUG|SLUGH|SNAI2|Zinc finger protein SNAI2|Neural crest transcription factor Slug|Protein snail homolog 2
  • Purity:
    >80%
  • Concentration:
    0,5 mg/ml
  • Source:
    E. coli
  • Biological activity:
    Standard
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB201878-100
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −20 °C
  • Formulation:
    pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,32% Tris HCl
  • Conjugation:
    Unconjugated
  • Tested applications:
    SDS-PAGE
  • Species:
    Human
  • Animal-free:
    N

Specifications