Human Recombinant Factor XII (from Wheat germ)
ABCAAB158410-10
: Abcam
:
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Wheat germ
- Species:Human
- Seq:1106
- Storage conditions:Store at −80 °C
- Biological activity:Standard
- Form:Liquid
- Animal-free:N
- Protein synonyms:Coagulation factor XII|Hageman factor|HAF|F12
- Protein/peptide name:Factor XII
- Formulation:pH: 8; Constituents: 0,79% Tris HCl, 0,31% Glutathione
- Shipping temperature:Dry Ice
- Tested applications:ELISA, WB
Recombinant Human Factor XII protein is a Human Full Length protein, in the 1 to 300 aa range, expressed in Wheat germ and suitable for ELISA, WB.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.