Order Entry
Export
Orders LinkContactUsLinkComponent
Human Recombinant ZIK1 (from E. coli)

Human Recombinant ZIK1 (from E. coli)

Catalog # ABCAAB172813-10
Supplier:   Abcam
Human Recombinant ZIK1 (from E. coli)
Catalog # ABCAAB172813-10
Supplier:  Abcam
CAS Number:  
Human Recombinant ZIK1 (from E. coli)
Human Recombinant ZIK1 (from E. coli)
CAS Number:  
Price
385,00 €
Log in for availability
1 * 10 µG
Product Details & Documents

Recombinant Human ZIK1 protein is a Human Full Length protein, in the 1 to 384 aa range, expressed in Escherichia coli, with >95% purity, <1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Form:
    Liquid
  • Protein/peptide name:
    ZIK1
  • Protein synonyms:
    ZNF762|Zinc finger protein interacting with ribonucleoprotein K|Zinc finger protein 762
  • Purity:
    >95%
  • Source:
    E. coli
  • Biological activity:
    Standard
  • Shipping temperature:
    Dry Ice
  • Cat. No.:
    ABCAAB172813-10
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    <1 EU/µg
  • Storage conditions:
    Store at −20 °C
  • Formulation:
    pH: 7,5, Constituents: 0,24% Tris
  • Conjugation:
    Unconjugated
  • Tested applications:
    HPLC, SDS-PAGE
  • Species:
    Human
  • Animal-free:
    N

Specifications