Human Recombinant Rab34 (from E. coli)
Catalog # ABCAAB115717-50
Supplier: Abcam
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:1202
- Storage conditions:Store at −20 °C
- Biological activity:Standard
- Form:Liquid
- Animal-free:N
- Protein synonyms:RAB39|RAH|Ras-related protein Rab-34|Ras-related protein Rab-39|Ras-related protein Rah
- Protein/peptide name:Rab34
- Purity:>90%
- Concentration:0,5 mg/ml
- Formulation:pH: 8, Constituents: 40% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,32% Tris HCl, 0,02% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
- Shipping temperature:20
- Tested applications:MS, SDS-PAGE
Specifications
About this item
Recombinant Human Rab34 protein is a Human Full Length protein, in the 1 to 259 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.