Human Recombinant beta Sarcoglycan (from Wheat germ)
Catalog # ABCAAB152676-10
Supplier: Abcam
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Wheat germ
- Species:Human
- Seq:1106
- Storage conditions:Store at −80 °C
- Biological activity:Standard
- Form:Liquid
- Animal-free:N
- Protein synonyms:Beta-sarcoglycan|Beta-SG|43 kDa dystrophin-associated glycoprotein|A3b|43DAG|SGCB
- Protein/peptide name:beta Sarcoglycan
- Formulation:pH: 8, Constituents: 0,79% Tris HCl, 0,31% Glutathione
- Shipping temperature:Dry Ice
- Tested applications:ELISA, SDS-PAGE, WB
Specifications
About this item
Recombinant Human beta Sarcoglycan protein is a Human Full Length protein, in the 1 to 318 aa range, expressed in Wheat germ and suitable for western blot, SDS-PAGE and ELISA. The predicted molecular weight of ab152676 protein is 60,7 kDa. - Save time and ensure accurate results - use our Human beta Sarcoglycan (SGCB) as a control
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.