Order Entry
Export
Orders LinkContactUsLinkComponent
Rat Recombinant PLBD2 (from E. coli)

Rat Recombinant PLBD2 (from E. coli)

Catalog # ABCAAB226421-100
Supplier:   Abcam
Rat Recombinant PLBD2 (from E. coli)
Catalog # ABCAAB226421-100
Supplier:  Abcam
CAS Number:  
Rat Recombinant PLBD2 (from E. coli)
Rat Recombinant PLBD2 (from E. coli)
CAS Number:  
Price
697,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Recombinant Rat PLBD2 protein (Tagged) is a Rat Full Length protein, in the 36 to 585 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Form:
    Liquid
  • Protein/peptide name:
    PLBD2
  • Protein synonyms:
    RDCR-0918-3|Plbd2|Putative phospholipase B-like 2|LAMA-like protein 2|Lamina ancestor homolog 2|Phospholipase B domain-containing protein 2
  • Purity:
    >90%
  • Source:
    E. coli
  • Biological activity:
    Standard
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB226421-100
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −20 °C
  • Formulation:
    pH: 7,2 - 7,4, Constituents: Tris buffer, 50% Glycerol (glycerin, glycerine)
  • Conjugation:
    Unconjugated
  • Tested applications:
    SDS-PAGE
  • Species:
    Rat
  • Animal-free:
    N

Specifications