Rat Recombinant IL-10 (from E. coli)
Recombinant Rat IL-10 protein (Animal Free) is a Rat Full Length protein, in the 19 to 178 aa range, expressed in Escherichia coli, with >95% purity, <1 EU/µg endotoxin level and suitable for SDS-PAGE.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Recombinant Rat IL-10 protein (Animal Free) is a Rat Full Length protein, in the 19 to 178 aa range, expressed in Escherichia coli, with >95% purity, <1 EU/µg endotoxin level and suitable for SDS-PAGE.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- Catalog No:
- ABCAAB256036-5
- ABCAAB256036-20
- ABCAAB256036-100
- ABCAAB256036-500
- ABCAAB256036-1
- Protein/peptide name:
- IL-10
- IL-10
- IL-10
- IL-10
- IL-10
- Protein synonyms:
- Il10|Interleukin-10|Cytokine synthesis inhibitory factor|CSIF
- Il10|Interleukin-10|Cytokine synthesis inhibitory factor|CSIF
- Il10|Interleukin-10|Cytokine synthesis inhibitory factor|CSIF
- Il10|Interleukin-10|Cytokine synthesis inhibitory factor|CSIF
- Il10|Interleukin-10|Cytokine synthesis inhibitory factor|CSIF
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Species:
- Rat
- Rat
- Rat
- Rat
- Rat
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Animal-free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin content:
- <1 EU/µg
- <1 EU/µg
- <1 EU/µg
- <1 EU/µg
- <1 EU/µg
- Biological activity:
- Standard
- Standard
- Standard
- Standard
- Standard
- Purity:
- >95%
- >95%
- >95%
- >95%
- >95%
- Form:
- Lyophilised
- Lyophilised
- Lyophilised
- Lyophilised
- Lyophilised
- Formulation:
- Constituents: 0,1% Trifluoroacetic acid
- Constituents: 0,1% Trifluoroacetic acid
- Constituents: 0,1% Trifluoroacetic acid
- Constituents: 0,1% Trifluoroacetic acid
- Constituents: 0,1% Trifluoroacetic acid
- Storage conditions:
- Store at −20 °C
- Store at −20 °C
- Store at −20 °C
- Store at −20 °C
- Store at −20 °C
- Shipping temperature:
- Ambient - Can ship with ice
- Ambient - Can ship with ice
- Ambient - Can ship with ice
- Ambient - Can ship with ice
- Ambient - Can ship with ice
- Tested applications:
- SDS-PAGE
- SDS-PAGE
- SDS-PAGE
- SDS-PAGE
- SDS-PAGE
Product Family Options
Specifications
Cat. No.ABCAAB256036-5Protein/peptide nameIL-10Protein synonymsIl10|Interleukin-10|Cytokine synthesis inhibitory factor|CSIFProtein/peptide typeRecombinantSpeciesRatSourceE. coliConjugationUnconjugatedAnimal-freeYEndotoxin content<1 EU/µgBiological activityStandardPurity>95%FormLyophilisedFormulationConstituents: 0,1% Trifluoroacetic acidStorage conditionsStore at −20 °CShipping temperatureAmbient - Can ship with iceTested applicationsSDS-PAGESpecifications
Cat. No.ABCAAB256036-20Protein/peptide nameIL-10Protein synonymsIl10|Interleukin-10|Cytokine synthesis inhibitory factor|CSIFProtein/peptide typeRecombinantSpeciesRatSourceE. coliConjugationUnconjugatedAnimal-freeYEndotoxin content<1 EU/µgBiological activityStandardPurity>95%FormLyophilisedFormulationConstituents: 0,1% Trifluoroacetic acidStorage conditionsStore at −20 °CShipping temperatureAmbient - Can ship with iceTested applicationsSDS-PAGESpecifications
Cat. No.ABCAAB256036-100Protein/peptide nameIL-10Protein synonymsIl10|Interleukin-10|Cytokine synthesis inhibitory factor|CSIFProtein/peptide typeRecombinantSpeciesRatSourceE. coliConjugationUnconjugatedAnimal-freeYEndotoxin content<1 EU/µgBiological activityStandardPurity>95%FormLyophilisedFormulationConstituents: 0,1% Trifluoroacetic acidStorage conditionsStore at −20 °CShipping temperatureAmbient - Can ship with iceTested applicationsSDS-PAGESpecifications
Cat. No.ABCAAB256036-500Protein/peptide nameIL-10Protein synonymsIl10|Interleukin-10|Cytokine synthesis inhibitory factor|CSIFProtein/peptide typeRecombinantSpeciesRatSourceE. coliConjugationUnconjugatedAnimal-freeYEndotoxin content<1 EU/µgBiological activityStandardPurity>95%FormLyophilisedFormulationConstituents: 0,1% Trifluoroacetic acidStorage conditionsStore at −20 °CShipping temperatureAmbient - Can ship with iceTested applicationsSDS-PAGESpecifications
Cat. No.ABCAAB256036-1Protein/peptide nameIL-10Protein synonymsIl10|Interleukin-10|Cytokine synthesis inhibitory factor|CSIFProtein/peptide typeRecombinantSpeciesRatSourceE. coliConjugationUnconjugatedAnimal-freeYEndotoxin content<1 EU/µgBiological activityStandardPurity>95%FormLyophilisedFormulationConstituents: 0,1% Trifluoroacetic acidStorage conditionsStore at −20 °CShipping temperatureAmbient - Can ship with iceTested applicationsSDS-PAGE



