Order Entry
Export
Orders LinkContactUsLinkComponent
 

Mouse Recombinant FGF8 (from E. coli)

Supplier:   Abcam
Mouse Recombinant FGF8 (from E. coli)
Supplier:  Abcam
Certificates
Mouse Recombinant FGF8 (from E. coli)

3 Options Available Below

Product Details & Documents

Recombinant human/mouse FGF8 protein (Active) is a Human Full Length protein, in the 23 to 215 aa range, expressed in Escherichia coli, with >95% purity, <0,05 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ABCAAB218083-5
  • ABCAAB218083-25
  • ABCAAB218083-100
  • Protein/peptide name:
  • FGF8
  • FGF8
  • FGF8
  • Protein synonyms:
  • AIGF|Fibroblast growth factor 8|FGF-8|Androgen-induced growth factor|Heparin-binding growth factor 8|HBGF-8
  • AIGF|Fibroblast growth factor 8|FGF-8|Androgen-induced growth factor|Heparin-binding growth factor 8|HBGF-8
  • AIGF|Fibroblast growth factor 8|FGF-8|Androgen-induced growth factor|Heparin-binding growth factor 8|HBGF-8
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Species:
  • Mouse
  • Mouse
  • Mouse
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Animal-free:
  • N
  • N
  • N
  • Endotoxin content:
  • <0,05 EU/µg
  • <0,05 EU/µg
  • <0,05 EU/µg
  • Biological activity:
  • Bioactive
  • Bioactive
  • Bioactive
  • Purity:
  • >95%
  • >95%
  • >95%
  • Form:
  • Lyophilised
  • Lyophilised
  • Lyophilised
  • Formulation:
  • Constituents: 0,29% Sodium chloride, 0,08% Sodium phosphate
  • Constituents: 0,29% Sodium chloride, 0,08% Sodium phosphate
  • Constituents: 0,29% Sodium chloride, 0,08% Sodium phosphate
  • Storage conditions:
  • Store at –20 °C
  • Store at –20 °C
  • Store at –20 °C
  • Shipping temperature:
  • 20
  • 20
  • 20
  • Tested applications:
  • FuncS, SDS-PAGE
  • FuncS, SDS-PAGE
  • FuncS, SDS-PAGE

Specifications

Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 5 µG
    1 * 5 µG - 267,00 €
    267,00 €
    1 * 5 µG
       267,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB218083-5
    Protein/peptide name
    FGF8
    Protein synonyms
    AIGF|Fibroblast growth factor 8|FGF-8|Androgen-induced growth factor|Heparin-binding growth factor 8|HBGF-8
    Protein/peptide type
    Recombinant
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Animal-free
    N
    Endotoxin content
    <0,05 EU/µg
    Biological activity
    Bioactive
    Purity
    >95%
    Form
    Lyophilised
    Formulation
    Constituents: 0,29% Sodium chloride, 0,08% Sodium phosphate
    Storage conditions
    Store at –20 °C
    Shipping temperature
    20
    Tested applications
    FuncS, SDS-PAGE
  • Log in for availability
    Log in for availability
    1 * 25 µG
    1 * 25 µG - 277,00 €
    277,00 €
    1 * 25 µG
       277,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB218083-25
    Protein/peptide name
    FGF8
    Protein synonyms
    AIGF|Fibroblast growth factor 8|FGF-8|Androgen-induced growth factor|Heparin-binding growth factor 8|HBGF-8
    Protein/peptide type
    Recombinant
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Animal-free
    N
    Endotoxin content
    <0,05 EU/µg
    Biological activity
    Bioactive
    Purity
    >95%
    Form
    Lyophilised
    Formulation
    Constituents: 0,29% Sodium chloride, 0,08% Sodium phosphate
    Storage conditions
    Store at –20 °C
    Shipping temperature
    20
    Tested applications
    FuncS, SDS-PAGE
  • Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG - 539,00 €
    539,00 €
    1 * 100 µG
       539,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB218083-100
    Protein/peptide name
    FGF8
    Protein synonyms
    AIGF|Fibroblast growth factor 8|FGF-8|Androgen-induced growth factor|Heparin-binding growth factor 8|HBGF-8
    Protein/peptide type
    Recombinant
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Animal-free
    N
    Endotoxin content
    <0,05 EU/µg
    Biological activity
    Bioactive
    Purity
    >95%
    Form
    Lyophilised
    Formulation
    Constituents: 0,29% Sodium chloride, 0,08% Sodium phosphate
    Storage conditions
    Store at –20 °C
    Shipping temperature
    20
    Tested applications
    FuncS, SDS-PAGE