Mouse Recombinant FGF-15 (from E. coli)
About this item
Recombinant Mouse FGF-15 protein is a Mouse Full Length protein, in the 26 to 218 aa range, expressed in E. coli, with >90% purity and suitable for SDS-PAGE.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
3 Options Available Below
Product Details & Documents
Recombinant Mouse FGF-15 protein is a Mouse Full Length protein, in the 26 to 218 aa range, expressed in E. coli, with >90% purity and suitable for SDS-PAGE.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- ABCAAB315426-20UG
- ABCAAB315426-100UG
- ABCAAB315426-1MG
- Protein/peptide name:
- FGF-15
- FGF-15
- FGF-15
- Protein synonyms:
- Fibroblast growth factor 15|FGF-15|Fgf15
- Fibroblast growth factor 15|FGF-15|Fgf15
- Fibroblast growth factor 15|FGF-15|Fgf15
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Species:
- Mouse
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Animal-free:
- N
- N
- N
- Purity:
- >90%
- >90%
- >90%
- Form:
- Lyophilised
- Lyophilised
- Lyophilised
- Formulation:
- pH: 7,4 - 8. Constituents: 6% Trehalose, 0,87% Ethylenediaminetetraacetic acid, disodium, dihydrate, 0,24% Tris, 0,05% dodecyl 2-(trimethylazaniumyl)ethyl phosphate.
- pH: 7,4 - 8. Constituents: 6% Trehalose, 0,87% Ethylenediaminetetraacetic acid, disodium, dihydrate, 0,24% Tris, 0,05% dodecyl 2-(trimethylazaniumyl)ethyl phosphate.
- pH: 7,4 - 8. Constituents: 6% Trehalose, 0,87% Ethylenediaminetetraacetic acid, disodium, dihydrate, 0,24% Tris, 0,05% dodecyl 2-(trimethylazaniumyl)ethyl phosphate.
- Storage conditions:
- Store at −20 °C
- Store at −20 °C
- Store at −20 °C
- Shipping temperature:
- 20
- 20
- 20
- Tested applications:
- SDS-PAGE
- SDS-PAGE
- SDS-PAGE
Specifications
Product Family Options
Product Information
- AvailabilityPrice
- Specifications:New Product
More
Specifications
Cat. No.ABCAAB315426-20UGProtein/peptide nameFGF-15Protein synonymsFibroblast growth factor 15|FGF-15|Fgf15Protein/peptide typeRecombinantSpeciesMouseSourceE. coliConjugationUnconjugatedAnimal-freeNPurity>90%FormLyophilisedFormulationpH: 7,4 - 8. Constituents: 6% Trehalose, 0,87% Ethylenediaminetetraacetic acid, disodium, dihydrate, 0,24% Tris, 0,05% dodecyl 2-(trimethylazaniumyl)ethyl phosphate.Storage conditionsStore at −20 °CShipping temperature20Tested applicationsSDS-PAGENew Product - Specifications:New Product
More
Specifications
Cat. No.ABCAAB315426-100UGProtein/peptide nameFGF-15Protein synonymsFibroblast growth factor 15|FGF-15|Fgf15Protein/peptide typeRecombinantSpeciesMouseSourceE. coliConjugationUnconjugatedAnimal-freeNPurity>90%FormLyophilisedFormulationpH: 7,4 - 8. Constituents: 6% Trehalose, 0,87% Ethylenediaminetetraacetic acid, disodium, dihydrate, 0,24% Tris, 0,05% dodecyl 2-(trimethylazaniumyl)ethyl phosphate.Storage conditionsStore at −20 °CShipping temperature20Tested applicationsSDS-PAGENew Product - Specifications:New Product
More
Specifications
Cat. No.ABCAAB315426-1MGProtein/peptide nameFGF-15Protein synonymsFibroblast growth factor 15|FGF-15|Fgf15Protein/peptide typeRecombinantSpeciesMouseSourceE. coliConjugationUnconjugatedAnimal-freeNPurity>90%FormLyophilisedFormulationpH: 7,4 - 8. Constituents: 6% Trehalose, 0,87% Ethylenediaminetetraacetic acid, disodium, dihydrate, 0,24% Tris, 0,05% dodecyl 2-(trimethylazaniumyl)ethyl phosphate.Storage conditionsStore at −20 °CShipping temperature20Tested applicationsSDS-PAGENew Product



