Human Recombinant BMP2 (from E. coli)
Supplier: Abcam
Certificates
About this item
Recombinant human BMP2 protein (Active) is a Human Full Length protein, in the 283 to 396 aa range, expressed in Escherichia coli, with >95% purity, <1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C
- Endotoxin content:<1 EU/µg
- Biological activity:Bioactive
- Form:Lyophilised
- Animal-free:N
- Protein synonyms:BMP2A|Bone morphogenetic protein 2|BMP-2|Bone morphogenetic protein 2A|BMP-2A
- Protein/peptide name:BMP2
- Purity:>95%
- Formulation:Constituents: 0,1% Trifluoroacetic acid
- Shipping temperature:Ambient - Can ship with ice
- Tested applications:FuncS, SDS-PAGE