Human Recombinant LIMD2 (from E. coli)
Recombinant human LIMD2 protein is a Human Full Length protein, in the 1 to 127 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- Form:Liquid
- Protein/peptide name:LIMD2
- Protein synonyms:SB143|LIMD2|LIM domain-containing protein 2
- Purity:>90%
- Concentration:0,5 mg/ml
- Source:E. coli
- Biological activity:Standard
- Shipping temperature:20
- Cat. No.:ABCAAB167895-100
- Protein/peptide type:Recombinant
- Storage conditions:Store at −20 °C
- Formulation:pH: 7,4, Constituents: 80% PBS, 20% Glycerol (glycerin, glycerine)
- Conjugation:Unconjugated
- Tested applications:MS, SDS-PAGE
- Species:Human
- Animal-free:N



