Order Entry
Export
Orders LinkContactUsLinkComponent
SAM ELISA Kit

SAM ELISA Kit

SAM ELISA Kit
Catalog # ANTIA326873-96
New Product
Supplier:   Antibodies.com
SAM ELISA Kit
Catalog # ANTIA326873-96
Supplier:  Antibodies.com
New Product
CAS Number:  
SAM ELISA Kit

SAM ELISA Kit

CAS Number:  
Price
545,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

SAM ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of SAM in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples.

SAM ELISA Kit employs the competitive enzyme immunoassay technique for the quantitative measurement of SAM in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples. The 96-well microtiter plate has been pre-coated with SAM antigen. During the incubation, SAM present in the samples or standards competes with the fixed amount of immobilised SAM for binding sites on the Biotinylated Anti-SAM Antibody. The more SAM present in a sample or standard, the less Biotinylated Anti-SAM Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-SAM Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of SAM present in each sample or standard. The concentration of SAM can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Delivery information: This is a Ready-To-Use ELISA kit that contains: Pre-Coated 96 Well Microplate, Lyophilised Standard, Sample Dilution Buffer, Biotinylated Detection Antibody (100X), Antibody Dilution Buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC Dilution Buffer, TMB Substrate, Stop Solution, Wash Buffer (25X), Plate Sealers, Foil Pouch, and Protocol Manual.

Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.

Specifications

Specifications

  • Description:
    SAM ELISA Kit
  • Primary antibody reactivity:
    Other
  • Sample type:
    Serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples
  • Sample volume:
    50 μl
  • Detection range:
    1,25 - 80 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Competitive
  • Time to Results:
    2 h
  • Cross reactivity:
    SAM ELISA Kit exhibits high specificity and excellent specificity for the detection of universal SAM. No significant cross-reactivity or interference between SAM and analogues was observed.
  • Cat. No.:
    ANTIA326873-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,75 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    SAM
  • Detection limit:
    Intra-assay CV: <8%, Inter-assay CV: <10%
  • Regulatory status:
    RUO

Specifications